Emulsion (main components are high polymeric and cyclic silicones); forms a protective film to coat split ends; conditioning agent for rinses and shampoos; glazing agent for hair.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Emulsion (main components are high polymeric and cyclic silicones); forms a protective film to coat split ends; conditioning agent for rinses and shampoos; glazing agent for hair.
Solubility
Soluble in water 1.759 mg/L @ 25°C.
Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
RUO – Research Use Only
M Calas; G Cordina; J Bompart; M Ben Bari; T Jei, M L Ancelin; H Vial. Antimalarial activity of molecules interfering with Plasmodium falciparum phospholipid metabolism. Structure-activity relationship analysis. Journal of Medicinal Chemistry. 1997, 40 (22), 3557-3566.
Juan C López-Montilla; Samir Pandey; Dinesh O Shah; Oscar D Crisalle. Removal of non-ionic organic pollutants from water via liquid-liquid extraction. Water Research. 2005, 39 (9), 1907-1913.