Thermo Scientific Chemicals

3-(Diphenylphosphino)propylamine, 97+%, Thermo Scientific Chemicals

Catalog number: H60672.03
1 g, Each
Thermo Scientific Chemicals

3-(Diphenylphosphino)propylamine, 97+%, Thermo Scientific Chemicals

Catalog number: H60672.03
1 g, Each
Quantity
Catalog number: H60672.03
also known as H60672-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
16605-03-1
IUPAC Name
(3-aminopropyl)diphenylphosphane
Molecular Formula
C15H18NP
InChI Key
DQZWMOWSTWWMPP-UHFFFAOYSA-N
SMILES
NCCCP(C1=CC=CC=C1)C1=CC=CC=C1
Form
Liquid
Assay by NMR
>97.0%
Appearance (Color)
Colorless to pale yellow

Description

3-(Diphenylphosphino)propylamine is used as a paharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-(Diphenylphosphino)propylamine is used as a paharmaceutical.

Solubility
Not miscible or difficult to mix in water.

Notes
Air sensitive. Store under inert gas. Store away from oxidizing agents, air, acids.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text