Potassium cyclopropyltrifluoroborate, tech. 90%, Thermo Scientific Chemicals
Potassium cyclopropyltrifluoroborate, tech. 90%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Potassium cyclopropyltrifluoroborate, tech. 90%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H60673.03
also known as H60673-03
Price (USD)/ Each
87.70
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
87.70
Add to cart
Potassium cyclopropyltrifluoroborate, tech. 90%, Thermo Scientific Chemicals
Catalog numberH60673.03
Price (USD)/ Each
87.70
-
Add to cart
Chemical Identifiers
CAS1065010-87-8
IUPAC Namepotassium cyclopropyltrifluoroboranuide
Molecular FormulaC3H5BF3K
InChI KeyCFMLURFHOSOXRC-UHFFFAOYSA-N
SMILES[K+].F[B-](F)(F)C1CC1
View more
SpecificationsSpecification SheetSpecification Sheet
Proton NMR88.0% min
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only