Thermo Scientific Chemicals

(S)-N-Boc-2-hydroxymethylmorpholine, 98%, Thermo Scientific Chemicals

Catalog number: H60680.03
1 g, Each
Thermo Scientific Chemicals

(S)-N-Boc-2-hydroxymethylmorpholine, 98%, Thermo Scientific Chemicals

Catalog number: H60680.03
1 g, Each
Quantity
Catalog number: H60680.03
also known as H60680-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
135065-76-8
IUPAC Name
tert-butyl 2-(hydroxymethyl)morpholine-4-carboxylate
Molecular Formula
C10H19NO4
InChI Key
FJYBLMJHXRWDAQ-UHFFFAOYNA-N
SMILES
CC(C)(C)OC(=O)N1CCOC(CO)C1
Assay (HPLC)
>97.5%

Description

(S)-N-Boc-2-hydroxymethylmorpholine is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(S)-N-Boc-2-hydroxymethylmorpholine is used as a pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Light sensitive. Store away from oxidizing agents, air, acids. Store in dark
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text