Thermo Scientific Chemicals

Imidazolidinyl urea, Thermo Scientific Chemicals

Catalog number: H60729.22
100 g, Each
Thermo Scientific Chemicals

Imidazolidinyl urea, Thermo Scientific Chemicals

Catalog number: H60729.22
100 g, Each
Quantity
Catalog number: H60729.22
also known as H60729-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
39236-46-9
IUPAC Name
3-[3-(hydroxymethyl)-2,5-dioxoimidazolidin-4-yl]-1-[({[3-(hydroxymethyl)-2,5-dioxoimidazolidin-4-yl]carbamoyl}amino)methyl]urea
Molecular Formula
C11H16N8O8
InChI Key
ZCTXEAQXZGPWFG-UHFFFAOYNA-N
SMILES
OCN1C(NC(=O)NCNC(=O)NC2N(CO)C(=O)NC2=O)C(=O)NC1=O
Appearance (Color)
White to pale yellow
Form
Solid
Assay (Titration)
>98%
Elemental Analysis
28%≥N≥26%
Loss on Drying
≤5.0%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text