10-Hydroxydecanoic acid, 96%, Thermo Scientific Chemicals
10-Hydroxydecanoic acid, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

10-Hydroxydecanoic acid, 96%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H60743.03
also known as H60743-03
Price (USD)/ Each
89.80
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
89.80
Add to cart
10-Hydroxydecanoic acid, 96%, Thermo Scientific Chemicals
Catalog numberH60743.03
Price (USD)/ Each
89.80
-
Add to cart
Chemical Identifiers
CAS1679-53-4
IUPAC Name10-hydroxydecanoic acid
Molecular FormulaC10H20O3
InChI KeyYJCJVMMDTBEITC-UHFFFAOYSA-N
SMILESOCCCCCCCCCC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)>95.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only