1-Propanesulfonyl chloride was used in synthesis of 1-alkyl-3-methylimidazolium alkanesulfonate ionic liquids.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-Propanesulfonyl chloride was used in synthesis of 1-alkyl-3-methylimidazolium alkanesulfonate ionic liquids.
Solubility
Decomposes in hot water.
Notes
Moisture sensitive. Store in dry temperature.
RUO – Research Use Only
General References:
- John M. Stewart.; Howard P. Cordts. Oxidative Ring Cleavage Reactions of Propylene Sulfide. J. Am. Chem. Soc. 1952, 75 (23),5880-5884.
- F. Bordwell.; William Hewett. Notes - Synthesis from Thiolacetates. I. Synthesis of Alkanesulfonyl Chlorides.J. Org. Chem. 1957, 22 (8),980-981.