Thermo Scientific Chemicals

Piperonyl butoxide, tech. 90%, Thermo Scientific Chemicals

Catalog number: H60771.AC
25 mL, Each
Thermo Scientific Chemicals

Piperonyl butoxide, tech. 90%, Thermo Scientific Chemicals

Catalog number: H60771.AC
25 mL, Each
Quantity
Catalog number: H60771.AC
also known as H60771-AC
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
51-03-6
Assay (GC)
>88.0%

Description

Piperonyl butoxide is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Piperonyl butoxide is used as a pharmaceutical intermediate.

Solubility
Soluble in benzene, ethanol and methanol. Not miscible or difficult to mix with water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text