Thermo Scientific Chemicals

4-Vinyl-1,3-dioxolan-2-one, 99%, Thermo Scientific Chemicals

Catalog number: H60822.18
50 g, Each
Thermo Scientific Chemicals

4-Vinyl-1,3-dioxolan-2-one, 99%, Thermo Scientific Chemicals

Catalog number: H60822.18
50 g, Each
Quantity
Catalog number: H60822.18
also known as H60822-18
Price (USD)
Have Questions?

Chemical Identifiers

CAS
4427-96-7
IUPAC Name
(4S)-4-ethenyl-1,3-dioxolan-2-one
Molecular Formula
C5H6O3
InChI Key
BJWMSGRKJIOCNR-BYPYZUCNSA-N
SMILES
C=C[C@H]1COC(=O)O1
Assay (GC)
>98.5%

Description

It is used to synthesize functional polymers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used to synthesize functional polymers.

Solubility
Insoluble in water.

Notes
Store at 2-8°C. Incompatible with oxidizing agents and heat.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text