Thermo Scientific Chemicals

tert-Butyl phenyl carbonate, 97%, Thermo Scientific Chemicals

Catalog number: H60867.22
100 g, Each
Thermo Scientific Chemicals

tert-Butyl phenyl carbonate, 97%, Thermo Scientific Chemicals

Catalog number: H60867.22
100 g, Each
Quantity
Catalog number: H60867.22
also known as H60867-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
6627-89-0
Appearance (Color)
Clear colorless
Assay (HPLC)
>97.5%
Form
Liquid

Description

tert-Butyl phenyl carbonate is used in determination of octanol-water partition coefficients by microemulsion electrokinetic chromatography. It is also used in the synthesis of 2-nitroindoles and as a reagent for mono-Boc protection of α,ω-diamines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
tert-Butyl phenyl carbonate is used in determination of octanol-water partition coefficients by microemulsion electrokinetic chromatography. It is also used in the synthesis of 2-nitroindoles and as a reagent for mono-Boc protection of α,ω-diamines.

Solubility
Slightly soluble in water.

Notes
Keep container tightly sealed. Store in refrigerated conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text