Thermo Scientific Chemicals

Nickel(II) trifluoromethanesulfonate, 96%, Thermo Scientific Chemicals

Catalog number: H60878.03
1 g, Each
Thermo Scientific Chemicals

Nickel(II) trifluoromethanesulfonate, 96%, Thermo Scientific Chemicals

Catalog number: H60878.03
1 g, Each
Quantity
Catalog number: H60878.03
also known as H60878-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
60871-84-3
Assay (Titration)
>95.0%

Description

It is used as a pharmaceutical intermediate and as OLED chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a pharmaceutical intermediate and as OLED chemicals.

Solubility
Partly soluble in water.

Notes
Hygroscopic. Incompatible with strong oxidizing agents.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text