Thermo Scientific Chemicals

Anthraquinone-2-carboxylic acid, 98%, Thermo Scientific Chemicals

Catalog number: H60929.03
1 g, Each
Thermo Scientific Chemicals

Anthraquinone-2-carboxylic acid, 98%, Thermo Scientific Chemicals

Catalog number: H60929.03
1 g, Each
Quantity
Catalog number: H60929.03
also known as H60929-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
117-78-2
IUPAC Name
9,10-dioxo-9,10-dihydroanthracene-2-carboxylic acid
Molecular Formula
C15H8O4
InChI Key
ASDLSKCKYGVMAI-UHFFFAOYSA-N
SMILES
OC(=O)C1=CC=C2C(=O)C3=CC=CC=C3C(=O)C2=C1
Assay (HPLC)
>97.5%

Description

Anthraquinone 2-carboxylic acid (AQ) as a novel electron shuttling mediator and attached electron relay for HRP. Efficient method for the generation of hydrogen peroxide by aerobic photooxidation of 2-propanol using anthraquinone-2-carboxylic acid and molecular oxygen is investigated.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Anthraquinone 2-carboxylic acid (AQ) as a novel electron shuttling mediator and attached electron relay for HRP. Efficient method for the generation of hydrogen peroxide by aerobic photooxidation of 2-propanol using anthraquinone-2-carboxylic acid and molecular oxygen is investigated.

Solubility
Soluble in hot Acetic acid(almost transparency).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text