It is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used as a pharmaceutical intermediate.
Solubility
Soluble in water.
Notes
Incompatible with strong oxidizing agents. Store under room temperature.
RUO – Research Use Only
Martin Newcomb.; Anne G. Glenn.; William G. Williams. Rate constants and Arrhenius functions for rearrangements of the 2,2-dimethyl-3-butenyl and (2,2-dimethylcyclopropyl)methyl radicals. J. Org. Chem. 1989, 54 (11),2675-2681.
Michael T. Crimmins.; Charlotte A. Carroll.; Angela J. Wells. Pinacol-type rearrangements of intramolecular photocycloadducts: Application of the 2,2-dimethyl-4-pentenoate protecting group. Tetrahedron Lett. 1998, 39 (39), 7005-7008.