It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.
Solubility
Slightly soluble in water.
Notes
Incompatible with strong oxidizing agents. Store under room temperature. Moisture sensitive.
RUO – Research Use Only
General References:
- Kyoko Nozaki, Naomasa Sato.; Hidemasa Takaya. Enantioselective synthesis of L- and D-isoserine via asymmetric hydrogenation of methyl N-phthaloyl-3-amino-2-oxopropanoate. Tetrahedron Lett. 1993, 4 (10),2179-2182.
- Jan Cz. Dobrowolski.; Michał H. Jamróz.; Robert Kołos.; Joanna E. Rode.; Michał K. Cyrańskie.; Joanna Sadlej. IR low-temperature matrix, X-ray and ab initio study on L-isoserine conformations. Phys. Chem. Chem. Phys. 2010, 12 (36),10818-10830.