Thermo Scientific Chemicals

Methylsuccinic acid, 99%, Thermo Scientific Chemicals

Catalog number: H60967.14
25 g, Each
Thermo Scientific Chemicals

Methylsuccinic acid, 99%, Thermo Scientific Chemicals

Catalog number: H60967.14
25 g, Each
Quantity
Catalog number: H60967.14
also known as H60967-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
498-21-5
IUPAC Name
2-methylbutanedioic acid
Molecular Formula
C5H8O4
InChI Key
WXUAQHNMJWJLTG-UHFFFAOYNA-N
SMILES
CC(CC(O)=O)C(O)=O
Assay (Titration)
>98.5%

Description

Methylsuccinic acid is used as a flux material and an important dyestuff intermediate. It is also used as an intermediate in medicine and pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methylsuccinic acid is used as a flux material and an important dyestuff intermediate. It is also used as an intermediate in medicine and pharmaceuticals.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents. Store under room temperature. Moisture sensitive.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text