4'-Bromo-2'-hydroxyacetophenone is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4′-Bromo-2′-hydroxyacetophenone is used as a pharmaceutical intermediate.
Solubility
Slightly soluble in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only
Stephen T. Ingalls.; Paul E. Minkler.; Charles L. Hoppel.; J. Eric Nordlander. Rhodium-catalyzed asymmetric arylation of α,β-unsaturated imines with arylstannanes. Catalytic asymmetric synthesis of allylic amines. Journal of Chromatography A. 1984, 299 365-376.
Andrey S. Klymchenko.; Yves Mely. 7-(2-Methoxycarbonylvinyl)-3-hydroxychromones: new dyes with red shifted dual emission. Tetrahedron Letters. 2004, 45 (45), 8391-8394.