It is an active pharmaceutical intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is an active pharmaceutical intermediates.
Solubility
Slightly soluble in water.
Notes
Store in cool dry ventilated place. Ensure proper ventilation.
RUO – Research Use Only
Moorthy S. S. Palanki.; Paul E. Erdman.; Leah M. Gayo-Fung.; Graziella I. Shevlin.; Robert W. Sullivan.; Mark E. Goldman.; Lynn J. Ransone.; Brydon L. Bennett.; Anthony M. Manning Mark J. Suto. Inhibitors of NF-κB and AP-1 Gene Expression: SAR Studies on the Pyrimidine Portion of 2-Chloro-4-trifluoromethylpyrimidine-5-[N-(3‘,5‘-bis(trifluoromethyl)phenyl)carboxamide]. J. Med. Chem. 2000, 43 (21),3995-4004.