Thermo Scientific Chemicals

Ethyl 2-chloro-4-methylpyrimidine-5-carboxylate, 95%, Thermo Scientific Chemicals

Catalog number: H61158.06
5 g, Each
Thermo Scientific Chemicals

Ethyl 2-chloro-4-methylpyrimidine-5-carboxylate, 95%, Thermo Scientific Chemicals

Catalog number: H61158.06
5 g, Each
Quantity
Catalog number: H61158.06
also known as H61158-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
188781-08-0
IUPAC Name
ethyl 2-chloro-4-methylpyrimidine-5-carboxylate
Molecular Formula
C8H9ClN2O2
InChI Key
XJNJPQSXRUJCIB-UHFFFAOYSA-N
SMILES
CCOC(=O)C1=CN=C(Cl)N=C1C
Assay (GC)
>94.0%

Description

It is an active pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an active pharmaceutical intermediates.

Solubility
Slightly soluble in water.

Notes
Store in cool dry ventilated place. Ensure proper ventilation.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text