This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
5-Fluoronicotinic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.
Solubility
Soluble in methanol.
Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
RUO – Research Use Only
G. F. Hawkins; Arthur Roe. The Preparation Of 5-Fluoronicotinic Acid And 5-Fluoronicotinamide1. J. Org. Chem. 1949, 14 (2), 328-332.
Edward Leete; George B. Bodem; Marissa F. Manuel. Formation Of 5-Fluoronicotine From 5-Fluoronicotinic Acid In Nicotiana Tabacum. Phytochemistry. 1971, 10 (11), 2687-2692.