Thermo Scientific Chemicals

2-(Trimethylsilyl)thiophene, 97%, Thermo Scientific Chemicals

Catalog number: H61170.06
5 g, Each
Thermo Scientific Chemicals

2-(Trimethylsilyl)thiophene, 97%, Thermo Scientific Chemicals

Catalog number: H61170.06
5 g, Each
Quantity
Catalog number: H61170.06
also known as H61170-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
18245-28-8
IUPAC Name
trimethyl(thiophen-2-yl)silane
Molecular Formula
C7H12SSi
InChI Key
OANGLGSZPSFVDY-UHFFFAOYSA-N
SMILES
C[Si](C)(C)C1=CC=CS1
Assay (GC)
>96.0%

Description

2-(Trimethylsilyl)thiophene is used to produce 2,5-bis-adamantan-1-yl-thiophene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(Trimethylsilyl)thiophene is used to produce 2,5-bis-adamantan-1-yl-thiophene.

Notes
Keep container tightly sealed. Store refrigerated conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text