4-Bromo-2-fluorophenylacetic acid, 98%, Thermo Scientific Chemicals
4-Bromo-2-fluorophenylacetic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Bromo-2-fluorophenylacetic acid, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number H61176.14
also known as H61176-14
Price (USD)/ Each
637.65
Online exclusive
708.00 
Save 70.35 (10%)
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
637.65
Online exclusive
708.00 
Save 70.35 (10%)
Add to cart
4-Bromo-2-fluorophenylacetic acid, 98%, Thermo Scientific Chemicals
Catalog numberH61176.14
Price (USD)/ Each
637.65
Online exclusive
708.00 
Save 70.35 (10%)
-
Add to cart
Chemical Identifiers
CAS114897-92-6
IUPAC Name2-(4-bromo-2-fluorophenyl)acetic acid
Molecular FormulaC8H6BrFO2
InChI KeyPNBIYFPZODYMOO-UHFFFAOYSA-N
SMILESOC(=O)CC1=C(F)C=C(Br)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only