Thermo Scientific Chemicals

4-Ethynylbenzonitrile, 97%, Thermo Scientific Chemicals

Catalog number: H61244.06
5 g, Each
Thermo Scientific Chemicals

4-Ethynylbenzonitrile, 97%, Thermo Scientific Chemicals

Catalog number: H61244.06
5 g, Each
Quantity
Catalog number: H61244.06
also known as H61244-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
3032-92-6
IUPAC Name
4-ethynylbenzonitrile
Molecular Formula
C9H5N
InChI Key
LAGNMUUUMQJXBF-UHFFFAOYSA-N
SMILES
C#CC1=CC=C(C=C1)C#N
Assay from Supplier's CofA
≥96.0% (GC)
Appearance (Color)
White to dark brown
Form
Powder or crystals

Description

4-Ethynylbenzonitrile is used as a synthetic fragment and as a test compound for cross-coupling reactions. It is used in the study of hydrogen bond formation in multifunctional molecules due to the presence of four hydrogen bonding sites. It is also involved in the preparation of 4-[(trimethylsilyl)ethynyl]benzonitrile.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Ethynylbenzonitrile is used as a synthetic fragment and as a test compound for cross-coupling reactions. It is used in the study of hydrogen bond formation in multifunctional molecules due to the presence of four hydrogen bonding sites. It is also involved in the preparation of 4-[(trimethylsilyl)ethynyl]benzonitrile.

Solubility
Slightly soluble in water.

Notes
Incompatible with strong oxidizing agents, acids and organic materials.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text