Thermo Scientific Chemicals

Trimethylsilanol, 95%, Thermo Scientific Chemicals

Catalog number: H61276.14
25 g, Each
Thermo Scientific Chemicals

Trimethylsilanol, 95%, Thermo Scientific Chemicals

Catalog number: H61276.14
25 g, Each
Quantity
Catalog number: H61276.14
also known as H61276-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1066-40-6
Assay (GC)
>94.0%

Description

Trimethylsilanol used as a hydrophobic coat on silicate surfaces. Used as an as an antimicrobial agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Trimethylsilanol used as a hydrophobic coat on silicate surfaces. Used as an as an antimicrobial agent.

Solubility
Soluble in water (1.00 mg/ml).

Notes
Store in cool, dry place in tightly closed container. With good ventilation. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text