Thermo Scientific Chemicals

2-Hydrazino-4-(trifluoromethyl)pyrimidine, 97%, Thermo Scientific Chemicals

Catalog number: H61287.03
1 g, Each
Thermo Scientific Chemicals

2-Hydrazino-4-(trifluoromethyl)pyrimidine, 97%, Thermo Scientific Chemicals

Catalog number: H61287.03
1 g, Each
Quantity
Catalog number: H61287.03
also known as H61287-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
197305-97-8
IUPAC Name
2-hydrazinyl-4-(trifluoromethyl)pyrimidine
Molecular Formula
C5H5F3N4
InChI Key
XQCAINVUBSAUBJ-UHFFFAOYSA-N
SMILES
NNC1=NC=CC(=N1)C(F)(F)F
Assay (unspecified)
>96.0%

Description

2-Hydrazino-4-(trifluoromethyl)pyrimidine may be used as a derivatization reagent for the simultaneous determination of testosterone (T) and dihydrotestosterone (DHT) in murine tissue and serum. Synthesis of organohydrazide chelate complexes of rhenium and technetium. Preparation of 1-(4-methoxyphenyl)ethanone [(4-trifluoromethyl)pyrimidine-2-yl]hydrazone (NAS-75).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Hydrazino-4-(trifluoromethyl)pyrimidine may be used as a derivatization reagent for the simultaneous determination of testosterone (T) and dihydrotestosterone (DHT) in murine tissue and serum. Synthesis of organohydrazide chelate complexes of rhenium and technetium. Preparation of 1-(4-methoxyphenyl)ethanone [(4-trifluoromethyl)pyrimidine-2-yl]hydrazone (NAS-75).

Solubility
Soluble in aqueous acid.

Notes
Store in cool, dry place in tightly closed container. With good ventilation. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text