Thermo Scientific Chemicals

Tetrafluoroterephthalonitrile, 98%, Thermo Scientific Chemicals

Catalog number: H61326.14
25 g, Each
Thermo Scientific Chemicals

Tetrafluoroterephthalonitrile, 98%, Thermo Scientific Chemicals

Catalog number: H61326.14
25 g, Each
Quantity
Catalog number: H61326.14
also known as H61326-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1835-49-0
IUPAC Name
tetrafluorobenzene-1,4-dicarbonitrile
Molecular Formula
C8F4N2
InChI Key
PCRSJGWFEMHHEW-UHFFFAOYSA-N
SMILES
FC1=C(F)C(C#N)=C(F)C(F)=C1C#N
Appearance (Color)
White to yellow-green to pale brown
Assay (HPLC)
>97.5%
Form
Powder

Description

Tetrafluoroterephthalonitrile can be used in the synthesis of Polymers of Intrinsic Micro porosity (PIM). It is used to study ultraviolet (UV)-rearranged polymers of PIM-1 membranes for efficient separation of H2 and CO2.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tetrafluoroterephthalonitrile can be used in the synthesis of Polymers of Intrinsic Micro porosity (PIM). It is used to study ultraviolet (UV)-rearranged polymers of PIM-1 membranes for efficient separation of H2 and CO2.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text