Methyl 5-bromopyrimidine-2-carboxylate, 95%, Thermo Scientific Chemicals
Methyl 5-bromopyrimidine-2-carboxylate, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Methyl 5-bromopyrimidine-2-carboxylate, 95%, Thermo Scientific Chemicals

Have Questions?
Quantity:
1 g
Catalog number H61339.03
also known as H61339-03
Price (USD)/ Each
153.00
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
153.00
Add to cart
Methyl 5-bromopyrimidine-2-carboxylate, 95%, Thermo Scientific Chemicals
Catalog numberH61339.03
Price (USD)/ Each
153.00
-
Add to cart
Chemical Identifiers
CAS89581-38-4
IUPAC Namemethyl 5-bromopyrimidine-2-carboxylate
Molecular FormulaC6H5BrN2O2
InChI KeyXILAKTMDKMVJQV-UHFFFAOYSA-N
SMILESCOC(=O)C1=NC=C(Br)C=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Assay (GC)>94.0%
FormCrystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only