Thermo Scientific Chemicals

6-(Boc-amino)hexanoic acid, 95%, Thermo Scientific Chemicals

Catalog number: H61350.14
25 g, Each
Thermo Scientific Chemicals

6-(Boc-amino)hexanoic acid, 95%, Thermo Scientific Chemicals

Catalog number: H61350.14
25 g, Each
Quantity
Catalog number: H61350.14
also known as H61350-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
6404-29-1
IUPAC Name
6-{[(tert-butoxy)carbonyl]amino}hexanoic acid
Molecular Formula
C11H21NO4
InChI Key
RUFDYIJGNPVTAY-UHFFFAOYSA-N
SMILES
CC(C)(C)OC(=O)NCCCCCC(O)=O
Assay (HPLC)
>94.0%

Description

6-(Boc-amino)hexanoic acid is used in the preparation of esters of 6-aminohexanoic acid as antibacterial agents. EACA is reported to inhibit chymotrypsin, Factor VIIa, lysine carboxy peptidase, plasmin, and plasminogen activator.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-(Boc-amino)hexanoic acid is used in the preparation of esters of 6-aminohexanoic acid as antibacterial agents. EACA is reported to inhibit chymotrypsin, Factor VIIa, lysine carboxy peptidase, plasmin, and plasminogen activator.

Solubility
Soluble in chloroform and ethyl acetate. Slightly soluble in water.

Notes
Store at -20°C. Keep container tightly closed. With good ventilation. Protect form heat and store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text