2-Bromo-5-pyridinemethanol, 95%, Thermo Scientific Chemicals
2-Bromo-5-pyridinemethanol, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Bromo-5-pyridinemethanol, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number H61375.06
also known as H61375-06
Price (USD)/ Each
146.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
146.00
Add to cart
2-Bromo-5-pyridinemethanol, 95%, Thermo Scientific Chemicals
Catalog numberH61375.06
Price (USD)/ Each
146.00
-
Add to cart
Chemical Identifiers
CAS122306-01-8
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)>94.0%
2-Bromo-5-pyridinemethanol is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromo-5-pyridinemethanol is used as a pharmaceutical intermediate.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only