9-Bromo-1-nonanol is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
9-Bromo-1-nonanol is used as a pharmaceutical intermediate.
Solubility
Soluble in dichloromethane, ethyl acetate and hexane.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only
Satoko Washiro.; Masahiro Yoshizawa.; Hiromitsu Nakajima.; Hiroyuki Ohno . Highly ion conductive flexible films composed of network polymers based on polymerizable ionic liquids. Polymer. 2004, 45 (5), 1577-1582.
Nestor Carballeira.; Janice E. Thompson.; Eser Ayanoglu.; Carl Djerassi. Biosynthetic studies of marine lipids. 5. The biosynthesis of long-chain branched fatty acids in marine sponges. J. Org. Chem. 1986, 51 (14), 2751-2756.