3-Bromo-4-fluorobenzyl bromide, 97%, Thermo Scientific Chemicals
3-Bromo-4-fluorobenzyl bromide, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Bromo-4-fluorobenzyl bromide, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Catalog number H61399.22
also known as H61399-22
Price (USD)/ Each
343.00
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
343.00
Add to cart
3-Bromo-4-fluorobenzyl bromide, 97%, Thermo Scientific Chemicals
Catalog numberH61399.22
Price (USD)/ Each
343.00
-
Add to cart
Chemical Identifiers
CAS78239-71-1
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)>96.0%
3-Bromo-4-fluorobenzyl bromide is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Bromo-4-fluorobenzyl bromide is used as a pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store in cool, well ventilated area. Keep container tightly closed. Light Sensitive. Moisture sensitive. Store under Argon.
RUO – Research Use Only