2,6-Dimethylbenzyl alcohol, 98%, Thermo Scientific Chemicals
2,6-Dimethylbenzyl alcohol, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2,6-Dimethylbenzyl alcohol, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
250 mg
5 g
Catalog number H61409.03
also known as H61409-03
Price (USD)/ Each
112.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
132.00 
Save 19.35 (15%)
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
112.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
132.00 
Save 19.35 (15%)
Add to cart
2,6-Dimethylbenzyl alcohol, 98%, Thermo Scientific Chemicals
Catalog numberH61409.03
Price (USD)/ Each
112.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
132.00 
Save 19.35 (15%)
-
Add to cart
Chemical Identifiers
CAS62285-58-9
IUPAC Name(2,6-dimethylphenyl)methanol
Molecular FormulaC9H12O
InChI KeyJPEYJQDKTDVJSZ-UHFFFAOYSA-N
SMILESCC1=CC=CC(C)=C1CO
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only