Thermo Scientific Chemicals

3-Fluoro-4-iodoaniline, 98%, Thermo Scientific Chemicals

Catalog number: H61427.03
1 g, Each
Thermo Scientific Chemicals

3-Fluoro-4-iodoaniline, 98%, Thermo Scientific Chemicals

Catalog number: H61427.03
1 g, Each
Quantity
Catalog number: H61427.03
also known as H61427-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
656-66-6
IUPAC Name
3-fluoro-4-iodoaniline
Molecular Formula
C6H5FIN
InChI Key
KUVVJHBHRIXJKI-UHFFFAOYSA-N
SMILES
NC1=CC(F)=C(I)C=C1
Assay (HPLC)
>97.5%

Description

3-Fluoro-4-iodoaniline is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Fluoro-4-iodoaniline is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

Solubility
Slightly soluble in water (228.3 mg/L at 25°C).

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is sensitive to light.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text