Thermo Scientific Chemicals

Thiophene-3-carbonyl chloride, 97%, Thermo Scientific Chemicals

Catalog number: H61428.06
5 g, Each
Thermo Scientific Chemicals

Thiophene-3-carbonyl chloride, 97%, Thermo Scientific Chemicals

Catalog number: H61428.06
5 g, Each
Quantity
Catalog number: H61428.06
also known as H61428-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
41507-35-1
IUPAC Name
thiophene-3-carbonyl chloride
Molecular Formula
C5H3ClOS
InChI Key
QTWBEVAYYDZLQL-UHFFFAOYSA-N
SMILES
ClC(=O)C1=CSC=C1
Assay (GC)
>96.0%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text