6-Amino-3-chloro-1H-indazole is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
6-Amino-3-chloro-1H-indazole is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.
Solubility
Sparingly soluble in water (0.62 g/L at 25°C).
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only
Shengqiang Wang.; Ruiyun Guo.; Jingya Li.; Dapeng Zou.; Yangjie Wu.; Yusheng Wu. Efficient synthesis of 3-aryl-1H-indazol-5-amine by Pd-catalyzed Suzuki-Miyaura cross-coupling reaction under microwave-assisted conditions. Tetrahedron Letters. 2015, 56 (24), 3750-3753.