Thermo Scientific Chemicals

2,6-Dibromopyrazine, 95%, Thermo Scientific Chemicals

Catalog number: H61468.03
1 g, Each
Thermo Scientific Chemicals

2,6-Dibromopyrazine, 95%, Thermo Scientific Chemicals

Catalog number: H61468.03
1 g, Each
Quantity
Catalog number: H61468.03
also known as H61468-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
23229-25-6
IUPAC Name
2,6-dibromopyrazine
Molecular Formula
C4H2Br2N2
InChI Key
JXKQTRCEKQCAGH-UHFFFAOYSA-N
SMILES
BrC1=CN=CC(Br)=N1
Assay (GC)
>94.0%

Description

2,6-Dibromopyrazine is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,6-Dibromopyrazine is used as a pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Storage temperature 2 - 4°Cand Keep container tightly closed in a dry and well-ventilated place.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text