2,6-Dibromopyrazine is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2,6-Dibromopyrazine is used as a pharmaceutical intermediate.
Solubility
Slightly soluble in water.
Notes
Storage temperature 2 - 4°Cand Keep container tightly closed in a dry and well-ventilated place.
RUO – Research Use Only
General References:
- Kurt H. Schaaf; Paul E. Spoerri. Syntheses in the Pyrazine Series: The Proof of the Structure and the Reactions of 2,6-Dibromopyrazine. J. Am. Chem. Soc. 1949, 71(6), 2043-2047.
- George Karmas; Paul E. Spoerri. Syntheses in the Pyrazine Series: Nucleophilic Displacements on Difunctional Pyrazines. J. Am. Chem. Soc. 1957, 79(3), 680-684.