Thermo Scientific Chemicals

4-Fluoro-1,3-dioxolan-2-one, 98%, Thermo Scientific Chemicals

Catalog number: H61502.22
100 g, Each
Thermo Scientific Chemicals

4-Fluoro-1,3-dioxolan-2-one, 98%, Thermo Scientific Chemicals

Catalog number: H61502.22
100 g, Each
Quantity
Catalog number: H61502.22
also known as H61502-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
114435-02-8
IUPAC Name
4-fluoro-1,3-dioxolan-2-one
Molecular Formula
C3H3FO3
InChI Key
SBLRHMKNNHXPHG-UHFFFAOYNA-N
SMILES
FC1COC(=O)O1
Assay (GC)
98%

Description

4-Fluoro-1,3-dioxolan-2-one is used as an electrolyte additive in lithium ion batteries. It also enables the formation of thin, smooth and stable passive solid electrolyte interphase (SEI) layer, which increases the cycling efficiency and discharge capacity retention of the secondary battery. Further, it is used as an intermediate in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Fluoro-1,3-dioxolan-2-one is used as an electrolyte additive in lithium ion batteries. It also enables the formation of thin, smooth and stable passive solid electrolyte interphase (SEI) layer, which increases the cycling efficiency and discharge capacity retention of the secondary battery. Further, it is used as an intermediate in organic synthesis.

Solubility
Slightly miscible with water.

Notes
Incompatible with strong oxidizing agents and strong bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text