Thermo Scientific Chemicals

6-Heptyn-1-ol, 95%, Thermo Scientific Chemicals

Catalog number: H61509.06
5 g, Each
Thermo Scientific Chemicals

6-Heptyn-1-ol, 95%, Thermo Scientific Chemicals

Catalog number: H61509.06
5 g, Each
Quantity
Catalog number: H61509.06
also known as H61509-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
63478-76-2
Assay (GC)
>94.0%

Description

6-Heptyn-1-ol is used as a reagent in in the preparation of many cyclic compounds. It is also used as a Intermediate in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Heptyn-1-ol is used as a reagent in in the preparation of many cyclic compounds. It is also used as a Intermediate in organic synthesis.

Solubility
Soluble in chloroform, dichloromethane and methanol. Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text