Thermo Scientific Chemicals

3-Bromo-5-methylaniline, 98%, Thermo Scientific Chemicals

Catalog number: H61519.06
5 g, Each
Thermo Scientific Chemicals

3-Bromo-5-methylaniline, 98%, Thermo Scientific Chemicals

Catalog number: H61519.06
5 g, Each
Quantity
Catalog number: H61519.06
also known as H61519-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
74586-53-1
IUPAC Name
3-bromo-5-methylaniline
Molecular Formula
C7H8BrN
InChI Key
YIZRPAWCIFTHNA-UHFFFAOYSA-N
SMILES
CC1=CC(N)=CC(Br)=C1
Assay (HPLC)
>97.5%

Description

3-Bromo-5-methylaniline is used as pharmaceutical intermediates and chemical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Bromo-5-methylaniline is used as pharmaceutical intermediates and chemical intermediate.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text