Thermo Scientific Chemicals

3-Maleimidopropionic acid, 95%, Thermo Scientific Chemicals

Catalog number: H61536.03
1 g, Each
Thermo Scientific Chemicals

3-Maleimidopropionic acid, 95%, Thermo Scientific Chemicals

Catalog number: H61536.03
1 g, Each
Quantity
Catalog number: H61536.03
also known as H61536-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7423-55-4
Form
Powder
Appearance (Color)
White to pale yellow
Assay (HPLC)
>94.0%

Description

3-Maleimidopropionic acid is a sulfhydryl reactive heterobifunctional crosslinking reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Maleimidopropionic acid is a sulfhydryl reactive heterobifunctional crosslinking reagent.

Solubility
Soluble in methanol, dimethyl formamide, alcohol and dimethyl sulfoxide. Slightly soluble in water.

Notes
Moisture sensitive. Store in cool place.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text