Thermo Scientific Chemicals

Methyl 4-fluoro-3-nitrobenzoate, 98%, Thermo Scientific Chemicals

Catalog number: H61542.22
100 g, Each
Thermo Scientific Chemicals

Methyl 4-fluoro-3-nitrobenzoate, 98%, Thermo Scientific Chemicals

Catalog number: H61542.22
100 g, Each
Quantity
Catalog number: H61542.22
also known as H61542-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
329-59-9
IUPAC Name
methyl 4-fluoro-3-nitrobenzoate
Molecular Formula
C8H6FNO4
InChI Key
CNJJSTPBUHAEFH-UHFFFAOYSA-N
SMILES
COC(=O)C1=CC(=C(F)C=C1)[N+]([O-])=O
Assay (HPLC)
97.5% min

Description

Methyl 4-fluoro-3-nitrobenzoate is used to prepare dimethyl 3-nitro-3',4-oxydibenzoate by reacting with 3-hydroxy-benzoic acid methyl ester.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 4-fluoro-3-nitrobenzoate is used to prepare dimethyl 3-nitro-3′,4-oxydibenzoate by reacting with 3-hydroxy-benzoic acid methyl ester.

Solubility
Soluble in ethanol, ether and methanol. Insoluble in water.

Notes
Incompatible with oxidizing agents and strong bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text