Thermo Scientific Chemicals

N-Boc-L-threonine methyl ester, 95%, Thermo Scientific Chemicals

Catalog number: H61545.06
5 g, Each
Thermo Scientific Chemicals

N-Boc-L-threonine methyl ester, 95%, Thermo Scientific Chemicals

Catalog number: H61545.06
5 g, Each
Quantity
Catalog number: H61545.06
also known as H61545-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
79479-07-5
IUPAC Name
methyl (2S,3R)-2-{[(tert-butoxy)carbonyl]amino}-3-hydroxybutanoate
Molecular Formula
C10H19NO5
InChI Key
MZMWAPNVRMDIPS-RQJHMYQMSA-N
SMILES
COC(=O)[C@@H](NC(=O)OC(C)(C)C)[C@@H](C)O
Assay (GC)
>94.0%

Description

N-Boc-L-threonine methyl ester is used as pharmaceutical intermediates and chemical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Boc-L-threonine methyl ester is used as pharmaceutical intermediates and chemical intermediate.

Solubility
Slightly soluble in water.

Notes
Storage temperature 2 - 4°C. Keep container tightly closed in a dry and well-ventilated place.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text