4-Cyano-3-fluoropyridine, 95%, Thermo Scientific Chemicals
4-Cyano-3-fluoropyridine, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Cyano-3-fluoropyridine, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H61594.03
also known as H61594-03
Price (USD)/ Each
166.00
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
166.00
Add to cart
4-Cyano-3-fluoropyridine, 95%, Thermo Scientific Chemicals
Catalog numberH61594.03
Price (USD)/ Each
166.00
-
Add to cart
Chemical Identifiers
CAS113770-88-0
IUPAC Name3-fluoropyridine-4-carbonitrile
Molecular FormulaC6H3FN2
InChI KeyRFQHDTRSEWCXJP-UHFFFAOYSA-N
SMILESFC1=C(C=CN=C1)C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>94.0%
4-Cyano-3-fluoropyridine is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Cyano-3-fluoropyridine is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and heat.
RUO – Research Use Only