N-Methyl-N-phenyl-beta-alanine ethyl ester, 99%, Thermo Scientific Chemicals
N-Methyl-N-phenyl-beta-alanine ethyl ester, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N-Methyl-N-phenyl-beta-alanine ethyl ester, 99%, Thermo Scientific Chemicals

Have Questions?
Quantity:
25 g
Catalog number H61615.14
also known as H61615-14
Price (USD)/ Each
144.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
144.00
Add to cart
N-Methyl-N-phenyl-beta-alanine ethyl ester, 99%, Thermo Scientific Chemicals
Catalog numberH61615.14
Price (USD)/ Each
144.00
-
Add to cart
Chemical Identifiers
CAS2003-76-1
IUPAC Nameethyl 3-[methyl(phenyl)amino]propanoate
Molecular FormulaC12H17NO2
InChI KeyKIGKCEWLNGHFCS-UHFFFAOYSA-N
SMILESCCOC(=O)CCN(C)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale yellow to amber
Impurity content≤ 0.1% (N-Methylaniline)
FormLiquid
Impurity content≤0.1% (Ethyl acrylate)
Assay (GC)>98.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only