1-Bromo-3-chloro-2-fluorobenzene, 95%, Thermo Scientific Chemicals
1-Bromo-3-chloro-2-fluorobenzene, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1-Bromo-3-chloro-2-fluorobenzene, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number H61622.14
also known as H61622-14
Price (USD)/ Each
55.50
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
55.50
Add to cart
1-Bromo-3-chloro-2-fluorobenzene, 95%, Thermo Scientific Chemicals
Catalog numberH61622.14
Price (USD)/ Each
55.50
-
Add to cart
Chemical Identifiers
CAS144584-65-6
IUPAC Name1-bromo-3-chloro-2-fluorobenzene
Molecular FormulaC6H3BrClF
InChI KeyDRNWNTAANHEQMK-UHFFFAOYSA-N
SMILESFC1=C(Cl)C=CC=C1Br
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>94.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only