N-Boc-3-iodo-L-alanine methyl ester, 98%, Thermo Scientific Chemicals
N-Boc-3-iodo-L-alanine methyl ester, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N-Boc-3-iodo-L-alanine methyl ester, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H61638.03
also known as H61638-03
Price (USD)/ Each
63.60
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
63.60
Add to cart
N-Boc-3-iodo-L-alanine methyl ester, 98%, Thermo Scientific Chemicals
Catalog numberH61638.03
Price (USD)/ Each
63.60
-
Add to cart
Chemical Identifiers
CAS93267-04-0
IUPAC Namemethyl (2S)-2-{[(tert-butoxy)carbonyl]amino}-3-iodopropanoate
Molecular FormulaC9H16INO4
InChI KeyUGZBFCCHLUWCQI-LURJTMIESA-N
SMILESCOC(=O)[C@H](CI)NC(=O)OC(C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>97.5%
N-Boc-3-iodo-L-alanine methyl ester is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Boc-3-iodo-L-alanine methyl ester is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

Solubility
Soluble in water and 1% acetic acid.

Notes
Light sensitive. Store in dark. Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
RUO – Research Use Only
Christian Meyer; Markus Scherer; Hartmut Schönberg; Heinz Rüegger; Sandra Loss; Volker Gramlich; and Hansjörg Grützmacher. Coordination chemistry of phosphanyl amino acids: solid state and solution structures of neutral and cationic rhodium complexes. Dalton Trans. 2006, (1), 137-148.