6-Fluoroindole-3-carboxaldehyde is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
6-Fluoroindole-3-carboxaldehyde is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.
Solubility
Soluble in methanol.
Notes
Store in freezer(-20°C). Keep container tightly closed. Store in a cool, dry and well-ventilated place.
RUO – Research Use Only
Hirohisa Saga; Takumi Ogawa; Kosuke Kai; Hideyuki Suzuki; Yoshiyuki Ogata; Nozomu Sakurai; Daisuke Shibata; and Daisaku Ohta. Identification and Characterization of ANAC042, a Transcription Factor Family Gene Involved in the Regulation of Camalexin Biosynthesis in Arabidopsis. Molecular Plant-Microbe Interactions. 2012, 25(5), 684-696.
Ines C. Raacke; Uta von Rad; Martin J. Mueller; and Susanne Berger.Yeast Increases Resistance in Arabidopsis Against Pseudomonas syringae and Botrytis cinerea by Salicylic Acid-Dependent as Well as -Independent Mechanisms. Molecular Plant-Microbe Interactions. 2006, 19(10), 1138-1146.