3-Bromo-4-fluorophenol, 98%, Thermo Scientific Chemicals
3-Bromo-4-fluorophenol, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Bromo-4-fluorophenol, 98%, Thermo Scientific Chemicals

Have Questions?
Quantity:
5 g
Catalog number H61702.06
also known as H61702-06
Price (USD)/ Each
136.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
136.00
Add to cart
3-Bromo-4-fluorophenol, 98%, Thermo Scientific Chemicals
Catalog numberH61702.06
Price (USD)/ Each
136.00
-
Add to cart
Chemical Identifiers
CAS27407-11-0
IUPAC Name3-bromo-4-fluorophenol
Molecular FormulaC6H4BrFO
InChI KeyQWTULQLVGNZMLF-UHFFFAOYSA-N
SMILESOC1=CC(Br)=C(F)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only