2-Amino-6-methyl-3-nitropyridine used as a intermediate in organic synthesis and used in medicine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Amino-6-methyl-3-nitropyridine used as a intermediate in organic synthesis and used in medicine.
Solubility
Slightly soluble in water.
Notes
Store in a cool, dry and well-ventilated place. Keep container tightly closed container. Store away from oxidizing agent.
RUO – Research Use Only
General References:
- Malvinder P. Singh; Tomi Joseph; Surat Kumar; Yadagiri Bathini; J. William Lown. Synthesis and sequence-specific DNA binding of a topoisomerase inhibitory analog of Hoechst 33258 designed for altered base and sequence recognition. Chem. Res. Toxicol. 1992, 5(5), 597-607.
- N. N. Smolyar; Yu. M. Yutilov. Reduction of 2-amino-3- and -5-nitropyridine derivatives with hydrazine hydrate. Russian Journal of Organic Chemistry. 2009, 45(1), 115-118.