Thermo Scientific Chemicals

2-Methyl-5-nitropyridine, 95%, Thermo Scientific Chemicals

Catalog number: H61750.06
5 g, Each
Thermo Scientific Chemicals

2-Methyl-5-nitropyridine, 95%, Thermo Scientific Chemicals

Catalog number: H61750.06
5 g, Each
Quantity
Catalog number: H61750.06
also known as H61750-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
21203-68-9
IUPAC Name
2-methyl-5-nitropyridine
Molecular Formula
C6H6N2O2
InChI Key
USZINSZJSVMICC-UHFFFAOYSA-N
SMILES
CC1=NC=C(C=C1)[N+]([O-])=O
Assay (HPLC)
>94.0%

Description

2-Methyl-5-nitropyridine is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methyl-5-nitropyridine is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text